Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Difluorobenzene, 99%

$78.50 - $78.50

Chemical Identifiers

Chemical Name or Material 1,4-Difluorobenzene
CAS 540-36-3
Molecular Formula C6H4F2
Formula Weight 114.09
MDL Number MFCD00000344
UN Number UN1993
Beilstein 1904541
Merck Index 14,3148
Synonym p-Difluorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1621714

 
alfa aesar™
A1621714*
25g Each for $78.50

AAA1621722

 
alfa aesar™
A1621722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.09
Molecular Formula C6H4F2
Beilstein 1904541
RTECS Number CZ5658000
Quantity 25g
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
UN Number UN1993
Chemical Name or Material 1,4-Difluorobenzene
Health Hazard 2 GHS H Statement
H225-H319
Highly flammable liquid and vapour.
Causes serious eye irritation.
Flash Point 2°C (35°F)
MDL Number MFCD00000344
Boiling Point 88° to 89°C
Synonym p-Difluorobenzene
Melting Point −13°C
Assay 99%
Health Hazard 3 GHS P Statement
P210-P260-P233-P243-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
Take precautionary measures against static discharge.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 1 Danger
Odor Characteristic
CAS 540-36-3
Merck Index 14,3148
Safety and Handling

Danger

GHS P Statement
P210-P260-P233-P243-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
Take precautionary measures against static discharge.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H225-H319
Highly flammable liquid and vapour.
Causes serious eye irritation.