Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Difluoro-2-nitrobenzene, 98%

Chemical Identifiers

Chemical Name or Material 1,4-Difluoro-2-nitrobenzene
CAS 364-74-9
Molecular Formula C6H3F2NO2
Formula Weight 159.09
MDL Number MFCD00007054
UN Number UN2810
Beilstein 2210200
Synonym 2,5-Difluoronitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1180714

  SDS
alfa aesar™
A1180714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1180722

  SDS
alfa aesar™
A1180722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 159.09
Chemical Name or Material 1,4-Difluoro-2-nitrobenzene
Health Hazard 2 GHS H Statement
H331-H373-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number CZ5715000
Assay 98%
Beilstein 2210200
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Health Hazard 3 GHS P Statement
P210-P260-P261-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 2,5-Difluoronitrobenzene
Molecular Formula C6H3F2NO2
CAS 364-74-9
Flash Point 90°C (194°F)
Quantity 25g
Melting Point −12°C
Boiling Point 206° to 207°C
MDL Number MFCD00007054
Health Hazard 1 Danger
UN Number UN2810
Safety and Handling

Danger

GHS P Statement
P210-P260-P261-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H373-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.