missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Difluoro-2-iodobenzene, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2689006

 View more versions of this product

Catalog No. AAH2689006



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,4-Difluoro-2-iodobenzene
1.998
85°C (185°F)
1.563
5 g
2,5-difluoroiodobenzene, benzene,1,4-difluoro-2-iodo, 2,5-difluoro-1-iodobenzene, benzene, 1,4-difluoro-2-iodo, pubchem4402, acmc-209fxx, 1,4-bis fluoranyl-2-iodanyl-benzene
FC1=CC=C(F)C(I)=C1
239.99
239.99
2265-92-1
178°C to 179°C
C6H3F2I
MFCD03094151
Light sensitive
WBYVNDUCUMNZPM-UHFFFAOYSA-N
1,4-difluoro-2-iodobenzene
223084
97%

Chemical Identifiers

2265-92-1
239.99
WBYVNDUCUMNZPM-UHFFFAOYSA-N
1,4-difluoro-2-iodobenzene
C6H3F2I
MFCD03094151
223084
FC1=CC=C(F)C(I)=C1
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only