Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Difluoro-2,5-dimethoxybenzene, 97%

$39.66 - $39.66

Chemical Identifiers

Chemical Name or Material 1,4-Difluoro-2,5-dimethoxybenzene
CAS 199866-90-5
Molecular Formula C8H8F2O2
Formula Weight 174.15g/mol
MDL Number MFCD00142553
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2540103

 
alfa aesar™
B25401-03
1g Each for $39.66
Description & Specifications

Specifications

Melting Point 119° to 122°C
Quantity 1g
Chemical Name or Material 1,4-Difluoro-2,5-dimethoxybenzene
CAS 199866-90-5
Assay 97%
Molecular Formula C8H8F2O2
Formula Weight 174.15g/mol
MDL Number MFCD00142553
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.