Learn More
1,4-Diethynylbenzene, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 391550050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 4-Diethynylbenzene | |
94.0 | |
94°C to 98°C | |
Authentic | |
Glass bottle | |
C6H4(C∽CH)2 | |
5 g | |
MVLGANVFCMOJHR-UHFFFAOYSA-N | |
1,4-diethynylbenzene | |
120463 | |
95% |
935-14-8 | |
100.0 | |
Brown-Orange or White | |
94% min. (GC) | |
C10H6 | |
MFCD00078375 | |
p-diethynylbenzene, benzene, 1,4-diethynyl, 1,4-diethynyl-benzene, benzene, diethynyl, benzene,1,4-diethynyl, benzene, p-diethynyl-7ci,8ci, 1,4-bis ethynyl benzene, acmc-209rm8, 4-05-00-01805 beilstein handbook reference | |
C#CC1=CC=C(C=C1)C#C | |
126.16 | |
126.16 | |
Crystalline Powder |
Chemical Identifiers
935-14-8 | |
126.16 | |
MVLGANVFCMOJHR-UHFFFAOYSA-N | |
120463 | |
C#CC1=CC=C(C=C1)C#C |
C10H6 | |
MFCD00078375 | |
p-diethynylbenzene, benzene, 1,4-diethynyl, 1,4-diethynyl-benzene, benzene, diethynyl, benzene,1,4-diethynyl, benzene, p-diethynyl-7ci,8ci, 1,4-bis ethynyl benzene, acmc-209rm8, 4-05-00-01805 beilstein handbook reference | |
1,4-diethynylbenzene |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO – Research Use Only