Please login to your online account to display your discounted pricing

1,4-Diethylpiperazine, 98%, ACROS Organics™

$114.93 - $114.93

Chemical Identifiers

Chemical Name or Material 1, 4-Diethylpiperazine
CAS 6483-50-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H18N2
Formula Weight 142.24
MDL Number MFCD00126900
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC244930250

  SDS
fisher scientific acros
244930250
Glass bottle 25g Each for $114.93
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 142.24
Quantity 25g
Color Clear colorless to yellow
Density 0.9000g/mL
CAS Max % 100.0
Chemical Name or Material 1, 4-Diethylpiperazine
Refractive Index 1.4530 to 1.4550 (20°C, 589nm)
Molecular Formula C8H18N2
MDL Number MFCD00126900
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
CAS 6483-50-7
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.