Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Diethoxybutane, 98%

Chemical Identifiers

Chemical Name or Material 1,4-Diethoxybutane
CAS 13344-00-8
Molecular Formula C8H17O2
Linear Formula CH3CH2O(CH2)4OCH2CH3
Formula Weight 146.23
MDL Number MFCD00048581
UN Number UN1993
Synonym 1,4-Butanediol diethyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2560706

  SDS
alfa aesar™
B2560706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2560714

  SDS
alfa aesar™
B2560714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 146.23
Linear Formula CH3CH2O(CH2)4OCH2CH3
Synonym 1,4-Butanediol diethyl ether
Molecular Formula C8H17O2
Quantity 5g
Flash Point 48°C (115°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Boiling Point 165°C
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
MDL Number MFCD00048581
CAS 13344-00-8
UN Number UN1993
Chemical Name or Material 1,4-Diethoxybutane
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.