Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Dicyclohexylbenzene, 99%

$38.18 - $332.49

Chemical Identifiers

Chemical Name or Material 1,4-Dicyclohexylbenzene
CAS 1087-02-1
Molecular Formula C18H26
Formula Weight 242.41
MDL Number MFCD00001452
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1870806

 
alfa aesar™
A1870806
5g Each for $38.18

AAA1870822

 
alfa aesar™
A1870822
100g Each for $332.49

AAA1870814

 
alfa aesar™
A1870814
25g Each for $103.76
Description & Specifications

Specifications

Formula Weight 242.41
Chemical Name or Material 1,4-Dicyclohexylbenzene
Assay 99%
Molecular Formula C18H26
MDL Number MFCD00001452
CAS 1087-02-1
Melting Point 103° to 105°C
Quantity 5g