missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Dicyanobutane, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 113832500
Specifications
1, 4-Dicyanobutane | |
163°C | |
Glass bottle | |
0.95 | |
295.0°C | |
1.0°C to 3.0°C | |
99% | |
98.5 | |
99% | |
NC(CH2)4CN | |
02, 653 | |
1,4-dicyanobutane, adipic acid dinitrile, adipic acid nitrile, adipinsaeuredinitril, adipodinitrile, adiponitrile, adipyldinitrile, nitrile adipico, tetramethylene cyanide, tetramethylene dicyanide | |
BTGRAWJCKBQKAO-UHFFFAOYSA-N | |
hexanedinitrile | |
8128 | |
Liquid |
0.9500g/mL | |
Authentic | |
1.4370 to 1.4390 | |
5.8 mPa.s (30°C) | |
Colorless to Yellow | |
250mL | |
111-69-3 | |
100.0 | |
C6H8N2 | |
MFCD00001975 | |
15, 152 | |
Solubility in water: 90g/L (20°C). Other solubilities: soluble in alcohol and chloroform | |
C(CCC#N)CC#N | |
108.14 | |
108.14 | |
99% |
Chemical Identifiers
111-69-3 | |
108.14 | |
BTGRAWJCKBQKAO-UHFFFAOYSA-N | |
8128 | |
C(CCC#N)CC#N |
C6H8N2 | |
MFCD00001975 | |
1,4-dicyanobutane, adipic acid dinitrile, adipic acid nitrile, adipinsaeuredinitril, adipodinitrile, adiponitrile, adipyldinitrile, nitrile adipico, tetramethylene cyanide, tetramethylene dicyanide | |
hexanedinitrile |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Harmful if inhaled.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and
GHS Signal Word: Danger
EINECSNumber : 203-896-3
RTECSNumber : AV2625000
TSCA : TSCA