Learn More
1,4-Dicyanobutane, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 113830010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 4-Dicyanobutane | |
111-69-3 | |
100.0 | |
Colorless to Yellow | |
295.0°C | |
Authentic | |
Glass bottle | |
1.4370 to 1.4390 | |
MFCD00001975 | |
02, 653 | |
15, 152 | |
Solubility in water: 90g/L (20°C). Other solubilities: soluble in alcohol and chloroform | |
C(CCC#N)CC#N | |
108.14 | |
5.8 mPa.s (30°C) | |
99% |
99% | |
98.5 | |
1.0°C to 3.0°C | |
0.9500g/mL | |
163°C | |
99% | |
C6H8N2 | |
NC(CH2)4CN | |
1 L | |
0.95 | |
adiponitrile, 1,4-dicyanobutane, adipodinitrile, adipyldinitrile, adipic acid dinitrile, tetramethylene dicyanide, nitrile adipico, tetramethylene cyanide, adipic acid nitrile, adipinsaeuredinitril | |
BTGRAWJCKBQKAO-UHFFFAOYSA-N | |
hexanedinitrile | |
8128 | |
108.14 | |
Liquid |
Chemical Identifiers
111-69-3 | |
108.14 | |
BTGRAWJCKBQKAO-UHFFFAOYSA-N | |
8128 | |
C(CCC#N)CC#N |
C6H8N2 | |
MFCD00001975 | |
adiponitrile, 1,4-dicyanobutane, adipodinitrile, adipyldinitrile, adipic acid dinitrile, tetramethylene dicyanide, nitrile adipico, tetramethylene cyanide, adipic acid nitrile, adipinsaeuredinitril | |
hexanedinitrile |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Harmful if inhaled.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and
GHS Signal Word: Danger
EINECSNumber : 203-896-3
RTECSNumber : AV2625000
TSCA : TSCA
RUO – Research Use Only