missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Dichlorophthalazine, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC155090050

 View more versions of this product

Catalog No. AC155090050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1, 4-Dichlorophthalazine
163.6°C to 163.8°C
4752-10-7
97.5
Glass bottle
98%
ODCNAEMHGMYADO-UHFFFAOYSA-N
1,4-dichlorophthalazine
78490
Powder or Crystalline Powder
White through Yellow
5g
Authentic
100.0
98%
C8H4Cl2N2
C1=CC=C2C(=C1)C(=NN=C2Cl)Cl
199.03
199.04

Chemical Identifiers

4752-10-7
199.03
78490
C8H4Cl2N2
ODCNAEMHGMYADO-UHFFFAOYSA-N
SDS Documents