missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Dichlorophthalazine, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC155090010

 View more versions of this product

Catalog No. AC155090010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1, 4-Dichlorophthalazine
163.6°C to 163.8°C
4767-03-7
97.5
Glass bottle
98%
PTBDIHRZYDMNKB-UHFFFAOYSA-N
3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
78501
Powder or Crystalline Powder
White through Yellow
1g
Authentic
100.0
98%
C5H10O4
CC(CO)(CO)C(=O)O
134.13
199.04

Chemical Identifiers

4767-03-7
134.13
78501
C5H10O4
PTBDIHRZYDMNKB-UHFFFAOYSA-N
SDS Documents