Please login to your online account to display your discounted pricing

1,4-Dichlorophthalazine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 4-Dichlorophthalazine
CAS 1041953
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H4Cl2N2
Formula Weight 199.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC155090010

 
thermo fisher scientific
AC155090010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC155090050

 
thermo fisher scientific
AC155090050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Crystalline Powder
Assay ≥97.5%
Formula Weight 199.04
Quantity 1g
Color White to yellow
Melting Point 163° to 167°C
CAS 1041953
CAS Max % 100.0
Chemical Name or Material 1, 4-Dichlorophthalazine
Molecular Formula C8H4Cl2N2
CAS Min % 97.5
Assay Percent Range 98%
Infrared Spectrum Authentic
Packaging Glass bottle