Please login to your online account to display your discounted pricing

1,4-Dichlorobutane, 99%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  216435000

 View more versions of this product

Catalog No. AC216435000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 127.01
Physical Form Liquid
Quantity 500mL
Color Clear
Beilstein 01, 119
EINECS Number 203-778-1
TSCA TSCA
Density 1.16g/mL
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.4530 to 1.4550 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Harmful to aquatic life with long lasting effects.
Flammable liquid and vapor.
Chemical Name or Material 1, 4-Dichlorobutane
Name Note 99+%
Water 0.05% max.
Assay Percent Range 99%
Molecular Formula C4H8Cl2
Linear Formula Cl(CH2)4Cl
MDL Number MFCD00001011
CAS 110-56-5
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 1, 4-Dichlorobutane
Name Note 99+%
CAS 110-56-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C4H8Cl2
Linear Formula Cl(CH2)4Cl
Formula Weight 127.01
MDL Number MFCD00001011
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Flammable liquid and vapor.

SDS & Safety Documents