missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Dichlorobutane, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 216431000

 View more versions of this product

Catalog No. AC216431000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1, 4-Dichlorobutane
110-56-5
100.0
1.1600g/mL
40°C
98.5% min. (GC)
C4H8Cl2
Cl(CH2)4Cl
100 mL
1.16
Solubility in water: 0.24% (20 c). Other solubilities: soluble in most common organic solvents
ClCCCCCl
127.01
1.46 mPa.s (20°C)
99%
99+%
98.5
-38°C
161°C to 163°C
Authentic
Glass bottle
1.4530 to 1.455
MFCD00001011
01, 119
butane, 1,4-dichloro, unii-8326ynm2b3, tetramethylene chloride, 1,4-dichlorobutane, butane,4-dichloro, tetramethylene dichloride, 1,4-bis chloranyl butane, ksc491k8b, 1,4-dichlorobutane, puriss gc
KJDRSWPQXHESDQ-UHFFFAOYSA-N
1,4-dichlorobutane
8059
127.01
Liquid

Chemical Identifiers

110-56-5
127.01
KJDRSWPQXHESDQ-UHFFFAOYSA-N
8059
ClCCCCCl
C4H8Cl2
MFCD00001011
butane, 1,4-dichloro, unii-8326ynm2b3, tetramethylene chloride, 1,4-dichlorobutane, butane,4-dichloro, tetramethylene dichloride, 1,4-bis chloranyl butane, ksc491k8b, 1,4-dichlorobutane, puriss gc
1,4-dichlorobutane
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Flammable liquid and vapor.

GHS P Statement:
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-778-1

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only