Learn More
1,4-Dichloro-2-nitrobenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1189636
Description
1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.
Solubility
Soluble in water, ethanol, ether, benzene, carbon disulfide. Slightly soluble in carbon tetrachloride.
Notes
Incompatible with strong oxidizing agents, strong bases.
Specifications
1,4-Dichloro-2-nitrobenzene | |
53°C to 57°C | |
266°C to 269°C | |
C6H3Cl2NO2 | |
500 g | |
778109 | |
Soluble in water,ethanol,ether,benzene,carbon disulfide. Slightly soluble in carbon tetrachloride. | |
[O-][N+](=O)C1=CC(Cl)=CC=C1Cl | |
192.00 | |
192 |
89-61-2 | |
1.6 | |
125°C (257°F) | |
MFCD00007074 | |
UN3077 | |
2,5-dichloronitrobenzene, nitro-p-dichlorobenzene, benzene, 1,4-dichloro-2-nitro, 1-nitro-2,5-dichlorobenzene, 2,5-dichlornitrobenzen, 2,5-dichloro-1-nitrobenzene, benzene, 2,5-dichloronitro, 2,5-dichloronitrobenzol, unii-iy18g50ff4, ccris 3096 | |
RZKKOBGFCAHLCZ-UHFFFAOYSA-N | |
1,4-dichloro-2-nitrobenzene | |
6977 | |
98% |
Chemical Identifiers
89-61-2 | |
192.00 | |
RZKKOBGFCAHLCZ-UHFFFAOYSA-N | |
6977 | |
[O-][N+](=O)C1=CC(Cl)=CC=C1Cl |
C6H3Cl2NO2 | |
MFCD00007074 | |
2,5-dichloronitrobenzene, nitro-p-dichlorobenzene, benzene, 1,4-dichloro-2-nitro, 1-nitro-2,5-dichlorobenzene, 2,5-dichlornitrobenzen, 2,5-dichloro-1-nitrobenzene, benzene, 2,5-dichloronitro, 2,5-dichloronitrobenzol, unii-iy18g50ff4, ccris 3096 | |
1,4-dichloro-2-nitrobenzene |
Safety and Handling
GHS H Statement
H341-H302-H319
Suspected of causing genetic defects.
Harmful if swallowed.
Causes serious eye irritation.
P264b-P270-P301+P312-P330-P501c
H302
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 201-923-3
RTECSNumber : CZ5260000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only