Please login to your online account to display your discounted pricing

1,4-Dichloro-2-iodobenzene, 98%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  175950500

 View more versions of this product

Catalog No. AC175950500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Color Clear colorless to brownish
Physical Form Liquid After Melting
Formula Weight 272.89
Quantity 50g
EINECS Number 249-774-3
Density 2.0200g/mL
TSCA TSCA
Beilstein 05, 221
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
MDL Number MFCD00001036
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.6450 to 1.6470 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Chemical Name or Material 1, 4-Dichloro-2-iodobenzene
CAS 29682-41-5
Assay Percent Range 98%
Linear Formula Cl2C6H3I
Molecular Formula C6 H3 Cl2 I
Name Note 98%

Specifications

Chemical Name or Material 1, 4-Dichloro-2-iodobenzene
Name Note 98%
CAS 29682-41-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6 H3 Cl2 I
Linear Formula Cl2C6H3I
Formula Weight 272.89
MDL Number MFCD00001036
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.

SDS & Safety Documents