missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Dichloro-2-iodobenzene, 98%, ACROS Organics™

Chemical Identifiers

CAS 29682-41-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C6 H3 Cl2 I
Linear Formula Cl2C6H3I
Formula Weight 272.89
MDL Number MFCD00001036
Chemical Name or Material 1, 4-Dichloro-2-iodobenzene, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC175950100 SDS Thermo Fisher Scientific
175950100
Glass bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

29682-41-5
100.0
C6 H3 Cl2 I
272.89
1, 4-Dichloro-2-iodobenzene, 98%
97.5
98%
Cl2C6H3I
MFCD00001036
≥97.5%

Specifications

2.0200g/mL
10g
97.5
Authentic
98%
Cl2C6H3I
MFCD00001036
1.6450 to 1.6470 (20°C, 589nm)
1, 4-Dichloro-2-iodobenzene, 98%
Clear colorless to brownish
29682-41-5
100.0
Liquid After Melting
C6 H3 Cl2 I
272.89
Glass bottle
05, 221
≥97.5%

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 249-774-3

TSCA : TSCA