Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Dichloro-2-iodobenzene, 98%

$39.49 - $551.47

Chemical Identifiers

Chemical Name or Material 1,4-Dichloro-2-iodobenzene
CAS 29682-41-5
Molecular Formula Cl2C6H3I
Formula Weight 272.9
MDL Number MFCD00001036
Beilstein 2245137
Synonym 2,5-Dichloroiodobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1417209

 
alfa aesar™
A1417209
10g Each for $39.49

AAA1417218

 
alfa aesar™
A1417218
50g Each for $130.03

AAA1417230

 
alfa aesar™
A1417230
250g Each for $551.47
Description & Specifications

Specifications

Formula Weight 272.9
MDL Number MFCD00001036
Quantity 10g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point 93°C (199°F)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Beilstein 2245137
Health Hazard 1 Warning
CAS 29682-41-5
Melting Point 20° to 21°C
Chemical Name or Material 1,4-Dichloro-2-iodobenzene
Synonym 2,5-Dichloroiodobenzene
Molecular Formula Cl2C6H3I
Boiling Point 255° to 257°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.