missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Dichloro-2-fluorobenzene, 99%, Thermo Scientific™
$276.16 - $276.16
Chemical Identifiers
CAS | 348-59-4 |
---|---|
Molecular Formula | C6H3Cl2F |
Molecular Weight (g/mol) | 164.99 |
MDL Number | MFCD00060656 |
InChI Key | BJQTYCQGIXZSNM-UHFFFAOYSA-N |
Synonym | 2,5-dichlorofluorobenzene, benzene, 1,4-dichloro-2-fluoro, 1,4-dichloro-2-fluoro-benzene, 2,5-dichloro-1-fluorobenzene, 1,4-dichlor-2-fluorbenzol, 2,5-dichloro-fluorobenzene, pubchem3454, acmc-1ae1b, 2,5-dichloro fluorobenzene, 1-fluor-2,5-dichlorbenzol |
PubChem CID | 136173 |
IUPAC Name | 1,4-dichloro-2-fluorobenzene |
SMILES | FC1=CC(Cl)=CC=C1Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2128214
|
Thermo Scientific Chemicals
B2128214 |
25 g |
Each for $276.16
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications1,4-Dichloro-2-fluorobenzene is used as organic synthesis intermediates
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
348-59-4 | |
164.99 | |
BJQTYCQGIXZSNM-UHFFFAOYSA-N | |
136173 | |
FC1=CC(Cl)=CC=C1Cl |
C6H3Cl2F | |
MFCD00060656 | |
2,5-dichlorofluorobenzene, benzene, 1,4-dichloro-2-fluoro, 1,4-dichloro-2-fluoro-benzene, 2,5-dichloro-1-fluorobenzene, 1,4-dichlor-2-fluorbenzol, 2,5-dichloro-fluorobenzene, pubchem3454, acmc-1ae1b, 2,5-dichloro fluorobenzene, 1-fluor-2,5-dichlorbenzol | |
1,4-dichloro-2-fluorobenzene |
Specifications
348-59-4 | |
1.383 | |
65°C (149°F) | |
1.5245 | |
25 g | |
2,5-dichlorofluorobenzene, benzene, 1,4-dichloro-2-fluoro, 1,4-dichloro-2-fluoro-benzene, 2,5-dichloro-1-fluorobenzene, 1,4-dichlor-2-fluorbenzol, 2,5-dichloro-fluorobenzene, pubchem3454, acmc-1ae1b, 2,5-dichloro fluorobenzene, 1-fluor-2,5-dichlorbenzol | |
BJQTYCQGIXZSNM-UHFFFAOYSA-N | |
1,4-dichloro-2-fluorobenzene | |
136173 | |
99% |
4°C | |
168°C | |
C6H3Cl2F | |
MFCD00060656 | |
2355590 | |
Difficult to mix. | |
FC1=CC(Cl)=CC=C1Cl | |
164.99 | |
165 | |
1,4-Dichloro-2-fluorobenzene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures