missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Dibromopentane, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0984222

 View more versions of this product

Catalog No. AAL0984222



Description

Description

1,4-Dibromopentane is used in the preparation of pharaoh ant trail pheromone stereoisomer, indolizidine alkaloid. It is also involved in the preparation 1,2-dimethyl-1-phenylcyclopentane and boc-L-2-amino-6-bromoheptanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dibromopentane is used in the preparation of pharaoh ant trail pheromone stereoisomer, indolizidine alkaloid. It is also involved in the preparation 1,2-dimethyl-1-phenylcyclopentane and boc-L-2-amino-6-bromoheptanoic acid.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

1,4-Dibromopentane
-34°C
102°C to 103°C (15 mmHg)
C5H10Br2
MFCD00000153
605295
Insoluble in water.
CC(CCCBr)Br
229.943
229.95
626-87-9
1.687
>110°C (230°F)
1.5085
100 g
pentane, 1,4-dibromo, 1,4-dibrompentan, pentane,4-dibromo, 1,4-dibromo-pentane, pubchem12546, 1,4-dibromopentane, hexanoic acid monoethyl ester
CNBFRBXEGGRSPL-UHFFFAOYSA-N
1,4-dibromopentane
79082
97%

Chemical Identifiers

626-87-9
229.943
CNBFRBXEGGRSPL-UHFFFAOYSA-N
79082
CC(CCCBr)Br
C5H10Br2
MFCD00000153
pentane, 1,4-dibromo, 1,4-dibrompentan, pentane,4-dibromo, 1,4-dibromo-pentane, pubchem12546, 1,4-dibromopentane, hexanoic acid monoethyl ester
1,4-dibromopentane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319

EINECSNumber : 210-967-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only