missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Dibromooctafluorobutane, 98%

$64.62 - $178.89

Chemical Identifiers

CAS 335-48-8
Molecular Formula C4Br2F8
Molecular Weight (g/mol) 359.839
MDL Number MFCD00153112
InChI Key RWWUGYJWSVESJC-UHFFFAOYSA-N
Synonym 1,4-dibromooctafluorobutane, 1,4-dibromoperfluorobutane, butane, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro, acmc-209i1o, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro-butane, butane,1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro, 1,4-bis bromanyl-1,1,2,2,3,3,4,4-octakis fluoranyl butane
PubChem CID 2736783
IUPAC Name 1,4-dibromo-1,1,2,2,3,3,4,4-octafluorobutane
SMILES C(C(C(F)(F)Br)(F)F)(C(F)(F)Br)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1673606 View Documents Alfa Aesar™
L1673606
5g Each for $64.62
 
AAL1673614 View Documents Alfa Aesar™
L1673614
25g Each for $178.89
 
Specifications

Chemical Identifiers

335-48-8
359.839
RWWUGYJWSVESJC-UHFFFAOYSA-N
2736783
C(C(C(F)(F)Br)(F)F)(C(F)(F)Br)(F)F
C4Br2F8
MFCD00153112
1,4-dibromooctafluorobutane, 1,4-dibromoperfluorobutane, butane, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro, acmc-209i1o, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro-butane, butane,1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro, 1,4-bis bromanyl-1,1,2,2,3,3,4,4-octakis fluoranyl butane
1,4-dibromo-1,1,2,2,3,3,4,4-octafluorobutane

Specifications

96°C to 98°C
335-48-8
Br(CF2)4Br
1777536
RWWUGYJWSVESJC-UHFFFAOYSA-N
1,4-dibromo-1,1,2,2,3,3,4,4-octafluorobutane
2736783
98%
5g
C4Br2F8
MFCD00153112
1,4-dibromooctafluorobutane, 1,4-dibromoperfluorobutane, butane, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro, acmc-209i1o, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro-butane, butane,1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro, 1,4-bis bromanyl-1,1,2,2,3,3,4,4-octakis fluoranyl butane
C(C(C(F)(F)Br)(F)F)(C(F)(F)Br)(F)F
359.839
359.84
1,4-Dibromooctafluorobutane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning