Learn More
1,4-Dibromobutane, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 161285000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 4-Dibromobutane | |
110-52-1 | |
100.0 | |
Brown-Yellow to Colorless | |
197°C | |
Authentic | |
Glass bottle | |
1.5176 to 1.5196 | |
MFCD00000261 | |
01, 120 | |
1.808 | |
Solubility in water: immiscible. Other solubilities: soluble in all usual organic solvents and alcohols,soluble in chloroform | |
C(CCBr)CBr | |
215.92 | |
215.92 | |
Liquid |
99% | |
98.5 | |
-20°C | |
1.8080g/mL | |
>112°C | |
98.5% min. (GC) | |
C4H8Br2 | |
Br(CH2)4Br | |
500 mL | |
13,313 | |
tetramethylene dibromide, butane, 1,4-dibromo, 1,4-butylene bromide, tetramethylene bromide, 1,4-dibrombutan, tetramethylenebromide, alpha,omega-dibromobutane, unii-g49phr6jfu, 1,4-dibrombutan german, 1,4-dibromo-butane | |
ULTHEAFYOOPTTB-UHFFFAOYSA-N | |
1,4-dibromobutane | |
8056 | |
99% |
Chemical Identifiers
110-52-1 | |
215.92 | |
ULTHEAFYOOPTTB-UHFFFAOYSA-N | |
8056 | |
C(CCBr)CBr |
C4H8Br2 | |
MFCD00000261 | |
tetramethylene dibromide, butane, 1,4-dibromo, 1,4-butylene bromide, tetramethylene bromide, 1,4-dibrombutan, tetramethylenebromide, alpha,omega-dibromobutane, unii-g49phr6jfu, 1,4-dibrombutan german, 1,4-dibromo-butane | |
1,4-dibromobutane |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Toxic if swallowed.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
EINECSNumber : 203-775-5
RTECSNumber : EJ7565000
TSCA : TSCA
RUO – Research Use Only