missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Dibromobutane, 98+%

Manufacturer:  Alfa Aesar™ A1166922*

 View more versions of this product

Catalog No. AAA1166922



Description

Description

Applications
1,4-Dibromobutane is used as an intermediate involved in the synthesis of active pharmaceutical ingredient and other organic compounds. It is also used in the investigation of metabolism of two halopropanes such as 1,3-dichloropropane and 2,2-dichloropropane. Further, it acts as a reagent to prepare diazadioxa oxovanadium(IV) macrocyclic complexes.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

1,4-Dibromobutane
194°C to 196°C
100g
≥98%
Br(CH2)4Br
1071199
ULTHEAFYOOPTTB-UHFFFAOYSA-N
1,4-dibromobutane
8056
131°C (267°F)
-20°C
110-52-1
C4H8Br2
MFCD00000261
Tetramethylene dibromide
C(CCBr)CBr
215.916
215.93

Chemical Identifiers

110-52-1
215.916
ULTHEAFYOOPTTB-UHFFFAOYSA-N
8056
C(CCBr)CBr
C4H8Br2
MFCD00000261
Tetramethylene dibromide
1,4-dibromobutane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : EJ7565000

TSCA : TSCA

SDS Documents