Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Dibromo-2-(trifluoromethoxy)benzene, JRD, 97%

$42.32 - $169.32

Chemical Identifiers

Chemical Name or Material 1,4-Dibromo-2-(trifluoromethoxy)benzene
CAS 175278-13-4
Name Note JRD
Molecular Formula C7H3Br2F3O
Formula Weight 319.9
MDL Number MFCD00179335
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3289603

  SDS
alfa aesar™
H3289603
1g Each for $42.32

AAH3289606

  SDS
alfa aesar™
H3289606
5g Each for $169.32
Description & Specifications

Specifications

Formula Weight 319.9
Chemical Name or Material 1,4-Dibromo-2-(trifluoromethoxy)benzene
Name Note JRD
Assay 97%
Quantity 1g
Molecular Formula C7H3Br2F3O
MDL Number MFCD00179335
CAS 175278-13-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.