missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Dibromo-2-ethylbenzene, 96%

$226.27 - $226.27

Chemical Identifiers

CAS 289039-53-8
Molecular Formula C8H8Br2
Formula Weight 263.96
MDL Number MFCD00672998
Chemical Name or Material 1,4-Dibromo-2-ethylbenzene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2544606 SDS Alfa Aesar™
B2544606
5g Each for $226.27
AAB2544603 SDS Alfa Aesar™
B2544603
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

289039-53-8
263.96
1,4-Dibromo-2-ethylbenzene
C8H8Br2
MFCD00672998

Specifications

289039-53-8
C8H8Br2
MFCD00672998
96%
5g
263.96
1,4-Dibromo-2-ethylbenzene

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning