Learn More
1,4-Dibromo-2,5-dimethoxybenzene, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1985818
Description
It is employed as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
1,4-Dibromo-2,5-dimethoxybenzene | |
144°C to 148°C | |
MFCD00025694 | |
1953103 | |
Insoluble in water. | |
COC1=CC(=C(C=C1Br)OC)Br | |
295.958 | |
295.96 |
2674-34-2 | |
C8H8Br2O2 | |
50 g | |
2,5-dibromo-1,4-dimethoxybenzene, 1,4-dibromo-2,5-dimethoxy-benzene, 2,5-dibromohydroquinone dimethyl ether, benzene, 1,4-dibromo-2,5-dimethoxy, pubchem16840, acmc-209gsv, maybridge1_005131, ksc496i0h, 2,5-dibromo-4-methoxyanisole, 1,4-dibromo-3,6-dimethoxybenzene | |
CHCLRVOURKGRSW-UHFFFAOYSA-N | |
1,4-dibromo-2,5-dimethoxybenzene | |
231240 | |
≥98% |
Chemical Identifiers
2674-34-2 | |
295.958 | |
CHCLRVOURKGRSW-UHFFFAOYSA-N | |
231240 | |
COC1=CC(=C(C=C1Br)OC)Br |
C8H8Br2O2 | |
MFCD00025694 | |
2,5-dibromo-1,4-dimethoxybenzene, 1,4-dibromo-2,5-dimethoxy-benzene, 2,5-dibromohydroquinone dimethyl ether, benzene, 1,4-dibromo-2,5-dimethoxy, pubchem16840, acmc-209gsv, maybridge1_005131, ksc496i0h, 2,5-dibromo-4-methoxyanisole, 1,4-dibromo-3,6-dimethoxybenzene | |
1,4-dibromo-2,5-dimethoxybenzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only