Please login to your online account to display your discounted pricing

1,4-Dibromo-2,5-difluorobenzene, 98%, Acros Organics


Quantity 25g

Manufacturer: thermo fisher scientific  249500250

 View more versions of this product

Catalog No. AC249500250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 271.89
Physical Form Adhering Crystals
Quantity 25g
Color White
Melting Point 59° to 63°C
EINECS Number 206-317-2
TSCA TSCA
CAS Max % 100.0
CAS Min % 97.5
Infrared Spectrum Authentic
Linear Formula Br2C6H2F2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Chemical Name or Material 1, 4-Dibromo-2, 5-difluorobenzene
CAS 327-51-5
Name Note 98%
Assay Percent Range 98%
MDL Number MFCD00000346
Molecular Formula C6H2Br2F2

Specifications

Chemical Name or Material 1, 4-Dibromo-2, 5-difluorobenzene
Name Note 98%
CAS 327-51-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H2Br2F2
Linear Formula Br2C6H2F2
Formula Weight 271.89
MDL Number MFCD00000346
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents