Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Dibenzyloxybenzene, 98%

$71.43 - $975.91

Chemical Identifiers

Chemical Name or Material 1,4-Dibenzyloxybenzene
CAS 621-91-0
Molecular Formula C20H18O2
Formula Weight 290.36
MDL Number MFCD00016875
Beilstein 2058196
Synonym Hydroquinone dibenzyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2494209

 
alfa aesar™
B2494209
10g Each for $71.43

AAB2494218

 
alfa aesar™
B2494218
50g Each for $247.82

AAB2494230

 
alfa aesar™
B2494230
250g Each for $975.91
Description & Specifications

Specifications

Formula Weight 290.36
Chemical Name or Material 1,4-Dibenzyloxybenzene
Melting Point 125° to 128°C
Assay 98%
Quantity 10g
Molecular Formula C20H18O2
Odor Odorless
MDL Number MFCD00016875
CAS 621-91-0
Beilstein 2058196
Synonym Hydroquinone dibenzyl ether