Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Dibenzoylbenzene, 99%

$96.28 - $355.59

Chemical Identifiers

Chemical Name or Material 1,4-Dibenzoylbenzene
CAS 3016-97-5
Molecular Formula C20H14O2
Formula Weight 286.33
MDL Number MFCD00014087
Beilstein 1880440
Synonym 4-Benzoylbenzophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1207006

  SDS
alfa aesar™
A1207006
5g Each for $96.28

AAA1207014

  SDS
alfa aesar™
A1207014
25g Each for $355.59
Description & Specifications

Specifications

Formula Weight 286.33
Synonym 4-Benzoylbenzophenone
Molecular Formula C20H14O2
MDL Number MFCD00014087
CAS 3016-97-5
Quantity 5g
Melting Point 161° to 165°C
Assay 99%
Beilstein 1880440
Chemical Name or Material 1,4-Dibenzoylbenzene