missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Diazabicyclo[2.2.2]octane, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC112470250

 View more versions of this product

Catalog No. AC112470250



Description

Description

Specifications

Specifications

1,4-Diazabicyclo[2.2.2]octane
62°C
Glass Bottle
20 °C
174.0°C
155.0°C to 160.0°C
97%
96.0
96% min. (GC)
MFCD00006689
15,9834
Solubility in water: 46g/100mL (26°C). Other solubilities: soluble in most common organic solvents
C1CN2CCN1CC2
112.17
112.17
97%
1.1400g/mL
Authentic
1.14
2.0 % max. (K.F.)
White to Yellow
25g
280-57-9
100.0
C6H12N2
01,1203; 02,99; 04,119; 05,176; 06,157; 07,86; 11,153; 12,155; 13,92; 15,109
1,4-diazabicyclo 2.2.2 octane, 1,4-ethylenepiperazine, dabco, dabco 33lv, dabco crystal, dabco s-25, teda, texacat td 100, triethylenediamine
IMNIMPAHZVJRPE-UHFFFAOYSA-N
1,4-diazabicyclo[2.2.2]octane
9237
Hygroscopic Crystals

Chemical Identifiers

280-57-9
112.17
IMNIMPAHZVJRPE-UHFFFAOYSA-N
9237
C1CN2CCN1CC2
C6H12N2
MFCD00006689
1,4-diazabicyclo 2.2.2 octane, 1,4-ethylenepiperazine, dabco, dabco 33lv, dabco crystal, dabco s-25, teda, texacat td 100, triethylenediamine
1,4-diazabicyclo[2.2.2]octane
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Danger

EINECSNumber : 205-999-9

RTECSNumber : HM0354200

TSCA : TSCA

SDS Documents