missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Diazabicyclo[2.2.2]octane, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC112470025
Specifications
1,4-Diazabicyclo[2.2.2]octane | |
62°C | |
Plastic bottle | |
20 °C | |
174.0°C | |
155.0°C to 160.0°C | |
97% | |
96.0 | |
96% min. (GC) | |
MFCD00006689 | |
15,9834 | |
Solubility in water: 46g/100mL (26°C). Other solubilities: soluble in most common organic solvents | |
C1CN2CCN1CC2 | |
112.17 | |
112.17 | |
97% |
1.1400g/mL | |
Authentic | |
1.14 | |
2.0 % max. (K.F.) | |
White to Yellow | |
2.5kg | |
280-57-9 | |
100.0 | |
C6H12N2 | |
01,1203; 02,99; 04,119; 05,176; 06,157; 07,86; 11,153; 12,155; 13,92; 15,109 | |
1,4-diazabicyclo 2.2.2 octane, 1,4-ethylenepiperazine, dabco, dabco 33lv, dabco crystal, dabco s-25, teda, texacat td 100, triethylenediamine | |
IMNIMPAHZVJRPE-UHFFFAOYSA-N | |
1,4-diazabicyclo[2.2.2]octane | |
9237 | |
Hygroscopic Crystals |
Chemical Identifiers
280-57-9 | |
112.17 | |
IMNIMPAHZVJRPE-UHFFFAOYSA-N | |
9237 | |
C1CN2CCN1CC2 |
C6H12N2 | |
MFCD00006689 | |
1,4-diazabicyclo 2.2.2 octane, 1,4-ethylenepiperazine, dabco, dabco 33lv, dabco crystal, dabco s-25, teda, texacat td 100, triethylenediamine | |
1,4-diazabicyclo[2.2.2]octane |
Safety and Handling
GHS H Statement
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Danger
EINECSNumber : 205-999-9
RTECSNumber : HM0354200
TSCA : TSCA