Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Diaminoanthraquinone, tech. 90%

$31.65 - $268.20

Chemical Identifiers

Chemical Name or Material 1,4-Diaminoanthraquinone
CAS 128-95-0
Purity Grade Technical Grade
Molecular Formula C14H10N2O2
Formula Weight 238.25
MDL Number MFCD00001224
Beilstein 2216556
Synonym C.I. 61100
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1915614

  SDS
alfa aesar™
A1915614
25g Each for $31.65

AAA1915622

  SDS
alfa aesar™
A1915622
100g Each for $88.15

AAA1915636

  SDS
alfa aesar™
A1915636
500g Each for $268.20
Description & Specifications

Specifications

Formula Weight 238.25
CAS 128-95-0
TSCA TSCA
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
RTECS Number CB6300000
MDL Number MFCD00001224
Molecular Formula C14H10N2O2
Odor Odorless
Assay 90%
Health Hazard 1 Danger
Synonym C.I. 61100
Chemical Name or Material 1,4-Diaminoanthraquinone
Quantity 25g
Melting Point 260° to 265°C
Beilstein 2216556
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Purity Grade Technical Grade

Applications
Use as the intermediate of synthetic dyes. Use anthraquinone reductive dyes and dispersal dyes and acid dyes and intermediates. Itself to disperse dyestuffs purple. 1,4-Diaminoanthraquinone oxidized with different metal atoms gives rise to conducting coordination polymers consisting of difunctional ligand molecules linked by metal atoms. 1, 4-diaminoanthraquinone has solar cell applications. A hybrid Si/poly (1, 4-diaminoanthraquinone) photoconductive diode for optical sensor applications. Acts as the sequential incorporation of the mediator in electrochemical sensing of glucose on a gold electrode.

Solubility
Soluble in benzene, pyridine, the difficulties, slightly soluble in hot aniline acetate, ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.