Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Diacryloylpiperazine, Electrophoresis Reagent, 97%

Chemical Identifiers

Chemical Name or Material 1,4-Diacryloylpiperazine
CAS 6342-17-2
Name Note Electrophoresis Reagent
Formula Weight 194.23
MDL Number MFCD00077661
Beilstein 745431
Synonym 1,4-Bis(acryloyl)piperazine; Diacroyl piperazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6642103

 
alfa aesar™
J6642103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ6642106

 
alfa aesar™
J6642106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ6642114

 
alfa aesar™
J6642114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.23
Melting Point 91° to 95°C
Chemical Name or Material 1,4-Diacryloylpiperazine
Physical Form Powder
Assay 97%
MDL Number MFCD00077661
CAS 6342-17-2
Beilstein 745431
Synonym 1,4-Bis(acryloyl)piperazine; Diacroyl piperazine
Name Note Electrophoresis Reagent
Color White to off-white
Quantity 1g