Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Diacryloylpiperazine, 97%

$213.96 - $753.38

Chemical Identifiers

Chemical Name or Material 1,4-Diacryloylpiperazine
CAS 6342-17-2
Molecular Formula C10H14N2O2
Formula Weight 194.23
MDL Number MFCD00077661
Beilstein 745431
Synonym 1,4-Bis(acryloyl)piperazine; Diacroyl piperazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1569406

  SDS
alfa aesar™
L1569406
5g Each for $213.96

AAL1569414

  SDS
alfa aesar™
L1569414
25g Each for $753.38
Description & Specifications

Specifications

Formula Weight 194.23
Chemical Name or Material 1,4-Diacryloylpiperazine
MDL Number MFCD00077661
Assay 97%
CAS 6342-17-2
Molecular Formula C10H14N2O2
Synonym 1,4-Bis(acryloyl)piperazine; Diacroyl piperazine
Beilstein 745431
Quantity 5g
RTECS Number UD6715000
Melting Point 93° to 96°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.