Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Diacetoxybutane, 98%

$71.43 - $230.89

Chemical Identifiers

Chemical Name or Material 1,4-Diacetoxybutane
CAS 628-67-1
Molecular Formula CH3CO2(CH2)4O2CCH3
Formula Weight 174.2
MDL Number MFCD00014984
Beilstein 1768757
Synonym 1,4-Butanediol diacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1697814

  SDS
alfa aesar™
A1697814
25g Each for $71.43

AAA1697822

  SDS
alfa aesar™
A1697822
100g Each for $230.89
Description & Specifications

Specifications

Formula Weight 174.2
Chemical Name or Material 1,4-Diacetoxybutane
Assay 98%
Molecular Formula CH3CO2(CH2)4O2CCH3
MDL Number MFCD00014984
CAS 628-67-1
Beilstein 1768757
Synonym 1,4-Butanediol diacetate
Boiling Point 230°C
Melting Point 12° to 15°C
Quantity 25g