Learn More
1,4-Diacetoxybenzene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2098730
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,4-Diacetoxybenzene | |
119°C to 122°C | |
MFCD00011643 | |
1912233 | |
AKOGNYJNGMLDOA-UHFFFAOYSA-N | |
(4-acetyloxyphenyl) acetate | |
71006 | |
98% |
1205-91-0 | |
C10H10O4 | |
250 g | |
1,4-diacetoxybenzene, hydroquinone diacetate, p-diacetoxybenzene, 1,4-benzenediol, diacetate, p-phenylene diacetate, hydroquinone, diacetate, 4-acetyloxy phenyl acetate, 1,4-phenylene diacetate, 1,4-benzenediol, 1,4-diacetate, benzene-1,4-diyl diacetate | |
CC(=O)OC1=CC=C(OC(C)=O)C=C1 | |
194.19 | |
194.19 |
Chemical Identifiers
1205-91-0 | |
194.19 | |
AKOGNYJNGMLDOA-UHFFFAOYSA-N | |
71006 | |
CC(=O)OC1=CC=C(OC(C)=O)C=C1 |
C10H10O4 | |
MFCD00011643 | |
1,4-diacetoxybenzene, hydroquinone diacetate, p-diacetoxybenzene, 1,4-benzenediol, diacetate, p-phenylene diacetate, hydroquinone, diacetate, 4-acetyloxy phenyl acetate, 1,4-phenylene diacetate, 1,4-benzenediol, 1,4-diacetate, benzene-1,4-diyl diacetate | |
(4-acetyloxyphenyl) acetate |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Warning
EINECSNumber : 214-887-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only