Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Diacetoxy-2-butyne, 97%

$64.34 - $64.34

Chemical Identifiers

Chemical Name or Material 1,4-Diacetoxy-2-butyne
CAS 1573-17-7
Molecular Formula C7H10O4
Linear Formula CH3CO2CH2C≡CCH2O2CCH3
Formula Weight 170.16
MDL Number MFCD00014982
Synonym 2-Butyne-1,4-diol diacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1818206

  SDS
alfa aesar™
A1818206
5g Each for $64.34
Description & Specifications

Specifications

Formula Weight 170.16
Boiling Point 116° to 118°C (8mmHg)
Melting Point 27° to 28°C
Linear Formula CH3CO2CH2C≡CCH2O2CCH3
Quantity 5g
MDL Number MFCD00014982
CAS 1573-17-7
Flash Point >110°C (230°F)
TSCA TSCA
Chemical Name or Material 1,4-Diacetoxy-2-butyne
Assay 97%
Molecular Formula C7H10O4
Synonym 2-Butyne-1,4-diol diacetate