missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Di-n-butoxy-2-nitrobenzene, 98%

$59.41 - $623.41

Chemical Identifiers

CAS 135-15-9
Molecular Formula C14H21NO4
Formula Weight 267.33
MDL Number MFCD00272266
Synonym 2,5-Di-n-butoxynitrobenzene
Chemical Name or Material 1,4-Di-n-butoxy-2-nitrobenzene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2470906 SDS Alfa Aesar™
B2470906
5g Each for $59.41
AAB2470914 SDS Alfa Aesar™
B2470914
25g Each for $200.11
AAB2470922 SDS Alfa Aesar™
B2470922
100g Each for $623.41
Specifications Description & Specifications

Chemical Identifiers

135-15-9
267.33
2,5-Di-n-butoxynitrobenzene
C14H21NO4
MFCD00272266
1,4-Di-n-butoxy-2-nitrobenzene

Specifications

135-15-9
C14H21NO4
MFCD00272266
1,4-Di-n-butoxy-2-nitrobenzene
5g
267.33
2,5-Di-n-butoxynitrobenzene
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning