missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Di(4-pyridyl)benzene, TCI America™

Manufacturer:  TCI America P15501G

 View more versions of this product

Catalog No. P15501G



Specifications

Specifications

1,4-Di(4-pyridyl)benzene
193°C
113682-56-7
C16H12N2
4,4′C-(1,4-Phenylene)bispyridine
C1=CC(=CC=C1C2=CC=NC=C2)C3=CC=NC=C3
232.286
232.29
White-Yellow
1g
≥95.0% (GC)
MFCD00445845
MAWKLXRVKVOYLR-UHFFFAOYSA-N
4-(4-pyridin-4-ylphenyl)pyridine
5250442
Crystal-Powder at 20°C

Chemical Identifiers

113682-56-7
232.286
MAWKLXRVKVOYLR-UHFFFAOYSA-N
5250442
C1=CC(=CC=C1C2=CC=NC=C2)C3=CC=NC=C3
C16H12N2
MFCD00445845
4,4′C-(1,4-Phenylene)bispyridine
4-(4-pyridin-4-ylphenyl)pyridine
Safety and Handling

Safety and Handling

TSCA : No

Documents