Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Cyclohexanedione mono-2,2-dimethyltrimethylene ketal, 95%

$442.99 - $442.99

Chemical Identifiers

Chemical Name or Material 1,4-Cyclohexanedione mono-2,2-dimethyltrimethylene ketal
CAS 69225-59-8
Molecular Formula C11H18O3
Formula Weight 198.26
MDL Number MFCD00006652
Beilstein 3541930
Synonym 3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0949014

  SDS
alfa aesar™
L0949014
25g Each for $442.99
Description & Specifications

Specifications

Formula Weight 198.26
Chemical Name or Material 1,4-Cyclohexanedione mono-2,2-dimethyltrimethylene ketal
Assay 95%
Molecular Formula C11H18O3
MDL Number MFCD00006652
CAS 69225-59-8
Synonym 3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one
Quantity 25g
Beilstein 3541930
Melting Point 46° to 50°C