missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,4-Cyclohexanedione mono-2,2-dimethyltrimethylene ketal, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L0949006
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1,4-Cyclohexanedione mono-2,2-dimethyltrimethylene ketal | |
46°C to 50°C | |
MFCD00006652 | |
3541930 | |
COKVDTKAWIFNTH-UHFFFAOYSA-N | |
3,3-dimethyl-1,5-dioxaspiro[5.5]undecan-9-one | |
587968 | |
95% |
69225-59-8 | |
C11H18O3 | |
5 g | |
3,3-dimethyl-1,5-dioxaspiro 5.5 undecan-9-one, 1,4-cyclohexanedione mono-2,2-dimethyltrimethylene ketal, 1,4-cyclohexanedione mono 2,2-dimethyltrimethylene ketal, 3,3-dimethyl-1,5-dioxa-spiro 5.5 undecan-9-one, 9,9-dimethyl-7,11-dioxaspiro 5.5 undecan-3-one, 1,5-dioxaspiro 5.5 undecan-9-one, 3,3-dimethyl, pubchem11226, maybridge1_004236, 1,4-cyclohexanedionemono-2,2-dimethyltrimethyleneketal | |
CC1(COC2(CCC(=O)CC2)OC1)C | |
198.262 | |
198.26 |
Chemical Identifiers
69225-59-8 | |
198.262 | |
COKVDTKAWIFNTH-UHFFFAOYSA-N | |
587968 | |
CC1(COC2(CCC(=O)CC2)OC1)C |
C11H18O3 | |
MFCD00006652 | |
3,3-dimethyl-1,5-dioxaspiro 5.5 undecan-9-one, 1,4-cyclohexanedione mono-2,2-dimethyltrimethylene ketal, 1,4-cyclohexanedione mono 2,2-dimethyltrimethylene ketal, 3,3-dimethyl-1,5-dioxa-spiro 5.5 undecan-9-one, 9,9-dimethyl-7,11-dioxaspiro 5.5 undecan-3-one, 1,5-dioxaspiro 5.5 undecan-9-one, 3,3-dimethyl, pubchem11226, maybridge1_004236, 1,4-cyclohexanedionemono-2,2-dimethyltrimethyleneketal | |
3,3-dimethyl-1,5-dioxaspiro[5.5]undecan-9-one |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 273-918-4
TSCA : No
Recommended Storage : Ambient temperatures