missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Cyclohexanedione, 98%

Manufacturer:  Alfa Aesar™ A1110036

 View more versions of this product

Catalog No. AAA1110036


 

Description

Description

Applications
1,4-Cyclohexanedione is used in the preparation of 1,4 benzoquinone and bromoorganics. It is also used to study the influence of visible light on the bromate-1,4-cyclohexanedione-ferroin oscillating reaction. It plays a vital role in pharmaceuticals, plant growth regulator and as a conducting material.

Solubility
Soluble in methanol and ethanol.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

1,4-Cyclohexanedione
98%
MFCD00001606
DCZFGQYXRKMVFG-UHFFFAOYSA-N
cyclohexane-1,4-dione
12511
112.13
77°C to 78°C
132°C (269°F)
637-88-7
C6H8O2
774152
C1CC(=O)CCC1=O
112.128
CHEBI:28286
132°C (20mmHg)
500g

Chemical Identifiers

637-88-7
112.128
DCZFGQYXRKMVFG-UHFFFAOYSA-N
CHEBI:28286
C6H8O2
MFCD00001606
12511
Safety and Handling

Safety and Handling

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents