Please login to your online account to display your discounted pricing

1,4-Cyclohexanedimethanol, Mixture of cis and trans 99%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: fisher scientific acros  154925000

 View more versions of this product

Catalog No. AC154925000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4880 to 1.4900 (20°C, 589nm)
Color Clear colorless
Quantity 500g
Chemical Name or Material 1, 4-Cyclohexanedimethanol
Name Note 99%
CAS 105-08-8
CAS Min % 98.5
CAS Max % 100.0
Physical Form Clear colorless viscous liquid after melting
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C8H16O2
Linear Formula C6H10(CH2OH)2
Formula Weight 144.21g/mol
MDL Number MFCD00001512
Synonym 1, 4-Bis(hydroxymethyl)cyclohexane
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number GU9800000
EINECS Number 203-268-9
TSCA TSCA

Specifications

Chemical Name or Material 1, 4-Cyclohexanedimethanol
Name Note 99%
CAS 105-08-8
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C8H16O2
Linear Formula C6H10(CH2OH)2
Formula Weight 144.21g/mol
MDL Number MFCD00001512
Synonym 1, 4-Bis(hydroxymethyl)cyclohexane
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.

SDS & Safety Documents