missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Cyclohexanedimethanol, cis + trans, 99%

$28.62 - $56.64

Chemical Identifiers

CAS 105-08-8
Molecular Formula C8H16O2
Formula Weight 144.21
MDL Number MFCD00001512
Beilstein 1902271
Synonym 1,4-Bis(hydroxymethyl)cyclohexane
Chemical Name or Material 1,4-Cyclohexanedimethanol, cis + trans
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1676936 Alfa Aesar™
A1676936
500g Each for $56.64
AAA1676922 Alfa Aesar™
A1676922
100g Each for $28.62
Specifications

Description

Applications
CHDM is a precursor to polyesters. It is one of the most important co-monomers for production of polyethylene terephthalate (PET), or polyethylene terephthalic ester (PETE).

Solubility
Miscible with water, glycolether and ketones.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Description & Specifications

Chemical Identifiers

105-08-8
144.21
1902271
1,4-Cyclohexanedimethanol, cis + trans
C8H16O2
MFCD00001512
1,4-Bis(hydroxymethyl)cyclohexane

Specifications

161°C (322°F)
≈30°C
105-08-8
144.21
1902271
1,4-Cyclohexanedimethanol, cis + trans
282° to 284°C
500g
C8H16O2
MFCD00001512
1,4-Bis(hydroxymethyl)cyclohexane
99%

Safety and Handling

RTECSNumber : GU9800000

TSCA : TSCA