missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Cyclohexanedicarboxylic acid, 99+%, mixture of cis and trans, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC406045000

 View more versions of this product

Catalog No. AC406045000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

1, 4-Cyclohexanedicarboxylic acid
1076-97-7
100.0
Powder
99+%
C8H12O4
MFCD00001465
White to off-white
500g
Mixture of cis and trans
99.0
Authentic
≥99.0%
Glass bottle
172.18
CHDA
164° to 168°C

Chemical Identifiers

1076-97-7
100.0
C8H12O4
MFCD00001465
1, 4-Cyclohexanedicarboxylic acid, Mixture of cis and trans
99.0
99+%
172.18
CHDA
≥99.0%