Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Cyclohexanebis(methylamine), cis + trans, 96%

Chemical Identifiers

Chemical Name or Material 1,4-Cyclohexanebis(methylamine)
CAS 2549-93-1
Name Note Cis + Trans
Molecular Formula C8H18N2
Formula Weight 142.25
MDL Number MFCD00039463
UN Number UN2922
Beilstein 2071383
Synonym 1,4-Bis(aminomethyl)cyclohexane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1340703

 
alfa aesar™
L1340703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1340706

 
alfa aesar™
L1340706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1340714

 
alfa aesar™
L1340714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 142.25
Chemical Name or Material 1,4-Cyclohexanebis(methylamine)
MDL Number MFCD00039463
Synonym 1,4-Bis(aminomethyl)cyclohexane
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 96%
Molecular Formula C8H18N2
UN Number UN2922
Beilstein 2071383
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
RTECS Number GU7000010
Flash Point 106°C (222°F)
Boiling Point 239° to 244°C
Quantity 1g
Name Note Cis + Trans
CAS 2549-93-1
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.