Learn More
1,4-Cyclohexanebis(methylamine), cis + trans, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1340706
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,4-Cyclohexanebis(methylamine) | |
2549-93-1 | |
239°C to 244°C | |
C8H18N2 | |
MFCD00039463 | |
UN2922 | |
Air Sensitive | |
OXIKYYJDTWKERT-UHFFFAOYSA-N | |
[4-(aminomethyl)cyclohexyl]methanamine | |
17354 | |
96% |
Cis + Trans | |
0.95 | |
106°C (222°F) | |
1.49 | |
5 g | |
2071383 | |
1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanedimethanamine, bamch, cis-1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanebis methylamine, cyclohexane-1,4-diyldimethanamine, trans-1,4-bis aminomethyl cyclohexane, 4-aminomethyl cyclohexyl methanamine, trans-1,4-cyclohexanedimethanamine, cyclohex-1,4-ylenebis methylamine | |
C1CC(CCC1CN)CN | |
142.246 | |
142.25 |
Chemical Identifiers
2549-93-1 | |
142.246 | |
OXIKYYJDTWKERT-UHFFFAOYSA-N | |
17354 | |
C1CC(CCC1CN)CN |
C8H18N2 | |
MFCD00039463 | |
1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanedimethanamine, bamch, cis-1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanebis methylamine, cyclohexane-1,4-diyldimethanamine, trans-1,4-bis aminomethyl cyclohexane, 4-aminomethyl cyclohexyl methanamine, trans-1,4-cyclohexanedimethanamine, cyclohex-1,4-ylenebis methylamine | |
[4-(aminomethyl)cyclohexyl]methanamine |
Safety and Handling
GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312-H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 219-840-6
RTECSNumber : GU7000010
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only