Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chlorophenylsulfonyl)-3,3-dimethyl-2-butanone, 98+%

$59.38 - $204.92

Chemical Identifiers

Chemical Name or Material 1-(4-Chlorophenylsulfonyl)-3,3-dimethyl-2-butanone
CAS 207974-06-9
Molecular Formula C12H15ClO3S
Formula Weight 274.77
MDL Number MFCD00082685
Synonym tert-Butyl 4-chlorophenylsulfonylmethyl ketone; 1-(4-Chlorobenzenesulfonyl)-3,3-dimethyl-2-butanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1142703

  SDS
alfa aesar™
L1142703
1g Each for $59.38

AAL1142706

  SDS
alfa aesar™
L1142706
5g Each for $204.92
Description & Specifications

Specifications

Formula Weight 274.77
MDL Number MFCD00082685
Chemical Name or Material 1-(4-Chlorophenylsulfonyl)-3,3-dimethyl-2-butanone
CAS 207974-06-9
Molecular Formula C12H15ClO3S
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
Quantity 1g
Melting Point 95° to 99°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Synonym tert-Butyl 4-chlorophenylsulfonylmethyl ketone; 1-(4-Chlorobenzenesulfonyl)-3,3-dimethyl-2-butanone
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.