missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Chlorophenylsulfonyl)-3,3-dimethyl-2-butanone, 98+%

Manufacturer:  Alfa Aesar™ L1142706

 View more versions of this product

Catalog No. AAL1142706



Specifications

Specifications

1-(4-Chlorophenylsulfonyl)-3,3-dimethyl-2-butanone
5g
C12H15ClO3S
1-4-chlorobenzenesulfonyl-3,3-dimethylbutan-2-one, 1-4-chlorophenyl sulfonyl-3,3-dimethylbutan-2-one, 1-4'-chlorobenzenesulfonyl-3,3-dimethylbutane-2-one, 1-4-chlorophenylsulfonyl-3,3-dimethyl-2-butanone, 1-4-chlorophenyl sulfonyl-3,3-dimethylbutan-2-one, 1-4-chlorophenyl sulfonyl-3,3-dimethyl-2-butanone, 1-4-chlorophenyl sulfonyl-3,3-dimethylbutan-2-one, 1-4'-chlorophenylsulfonyl-3,3-dimethylbutane-2-one, 1-4-chlorobenzenesulfonyl-3,3-dimethyl-2-butanone, 1-4-chlorophenyl sulfonyl-3,3-dimethyl-butan-2-one
CC(C)(C)C(=O)CS(=O)(=O)C1=CC=C(C=C1)Cl
274.759
274.77
95°C to 99°C
207974-06-9
MFCD00082685
XTIUXIMZIKBBSU-UHFFFAOYSA-N
1-(4-chlorophenyl)sulfonyl-3,3-dimethylbutan-2-one
545199
≥98%

Chemical Identifiers

207974-06-9
274.759
XTIUXIMZIKBBSU-UHFFFAOYSA-N
545199
CC(C)(C)C(=O)CS(=O)(=O)C1=CC=C(C=C1)Cl
C12H15ClO3S
MFCD00082685
1-4-chlorobenzenesulfonyl-3,3-dimethylbutan-2-one, 1-4-chlorophenyl sulfonyl-3,3-dimethylbutan-2-one, 1-4'-chlorobenzenesulfonyl-3,3-dimethylbutane-2-one, 1-4-chlorophenylsulfonyl-3,3-dimethyl-2-butanone, 1-4-chlorophenyl sulfonyl-3,3-dimethylbutan-2-one, 1-4-chlorophenyl sulfonyl-3,3-dimethyl-2-butanone, 1-4-chlorophenyl sulfonyl-3,3-dimethylbutan-2-one, 1-4'-chlorophenylsulfonyl-3,3-dimethylbutane-2-one, 1-4-chlorobenzenesulfonyl-3,3-dimethyl-2-butanone, 1-4-chlorophenyl sulfonyl-3,3-dimethyl-butan-2-one
1-(4-chlorophenyl)sulfonyl-3,3-dimethylbutan-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents